missing translation for 'onlineSavingsMsg'
Learn More

Fast Green FCF, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013631
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013631 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013631 Supplier Merck Emd Millipore Supplier No. 6872425MG

missing translation for 'validMainframeMsg'

Fast Green FCF is a synthetic organic dye, which belongs to the triphenylmethane group. It can be found in a wide range of applications in food and beverage industry. It can also be used in scientific research for the staining of proteins and identification of extracellular recording or injection sites in electrophysiological experiments. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 2353-45-9
Physical Form Neat
Linear Formula C37H34N2O10S3 Na2
MDL Number MFCD00013053
Melting Point 290°C
Synonym Food green 3
Type Analytical Standard
Quantity 25 mg
Formula Weight 808.85
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less