missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FARP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142075
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142075 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5142075 Supplier Invitrogen™ Supplier No. PA5142075
Only null left

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Functions as guanine nucleotide exchange factor for RAC1. May play a role in semaphorin signaling. Plays a role in the assembly and disassembly of dendritic filopodia, the formation of dendritic spines, regulation of dendrite length and ultimately the formation of synapses.

Specifications

Antigen FARP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FARP1
Gene Accession No. Q9Y4F1
Gene Alias CDEP; chondrocyte-derived ezrin-like protein; FARP1; FARP1-IT1; FERM, ARH/RhoGEF and pleckstrin domain protein 1; FERM, ARHGEF and pleckstrin domain-containing protein 1; FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived); FERM, RhoGEF and pleckstrin domain-containing protein 1; PH domain-containing family C member 2; pleckstrin homology domain-containing family C member 2; PLEKHC2; PPP1R75; protein phosphatase 1, regulatory subunit 75
Gene Symbols FARP1
Host Species Goat
Immunogen Peptide with sequence GEIEQRPTPGSRL-C (aa 2-14).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10160
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less