missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Farnesyl Acetate (mixture of isomers) 95.0+%, TCI America™
Supplier: TCI America F034725ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| 29548-30-9 | |
| 264.409 | |
| ZGIGZINMAOQWLX-NCZFFCEISA-N | |
| 638500 | |
| CC(=CCCC(=CCCC(=CCOC(=O)C)C)C)C |
| C17H28O2 | |
| MFCD00036516 | |
| farnesyl acetate, farnesol acetate, all-trans-farnesyl acetate, unii-d5zj1foc2i, trans,trans-farnesyl acetate, d5zj1foc2i, 3,7,11-trimethyl-2,6,10-dodecatrienyl acetate, 2e,6e-farnesyl acetate, 2,6,10-dodecatrien-1-ol, 3,7,11-trimethyl-, acetate, e,e, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate | |
| [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate |
Specifications
| Farnesyl Acetate (mixture of isomers) | |
| Yellow | |
| C17H28O2 | |
| MFCD00036516 | |
| ZGIGZINMAOQWLX-NCZFFCEISA-N | |
| [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate | |
| 638500 | |
| ≥95.0% (GC) |
| 29548-30-9 | |
| 140°C | |
| 25 mL | |
| farnesyl acetate, farnesol acetate, all-trans-farnesyl acetate, unii-d5zj1foc2i, trans,trans-farnesyl acetate, d5zj1foc2i, 3,7,11-trimethyl-2,6,10-dodecatrienyl acetate, 2e,6e-farnesyl acetate, 2,6,10-dodecatrien-1-ol, 3,7,11-trimethyl-, acetate, e,e, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate | |
| CC(=CCCC(=CCCC(=CCOC(=O)C)C)C)C | |
| 264.409 | |
| 264.41 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0762
TSCA : Yes