missing translation for 'onlineSavingsMsg'
Learn More

Farnesyl Acetate (mixture of isomers) 95.0+%, TCI America™

Catalog No. p-7129627
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03475ML 5 mL
F034725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03475ML Supplier TCI America Supplier No. F03475ML

Chemical Identifiers

CAS 29548-30-9
Molecular Formula C17H28O2
Molecular Weight (g/mol) 264.409
MDL Number MFCD00036516
InChI Key ZGIGZINMAOQWLX-NCZFFCEISA-N
Synonym farnesyl acetate, farnesol acetate, all-trans-farnesyl acetate, unii-d5zj1foc2i, trans,trans-farnesyl acetate, d5zj1foc2i, 3,7,11-trimethyl-2,6,10-dodecatrienyl acetate, 2e,6e-farnesyl acetate, 2,6,10-dodecatrien-1-ol, 3,7,11-trimethyl-, acetate, e,e, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
PubChem CID 638500
IUPAC Name [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate
SMILES CC(=CCCC(=CCCC(=CCOC(=O)C)C)C)C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 mL
Formula Weight 264.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Farnesyl Acetate (mixture of isomers)
EINECSNumber (2)-0762
TSCA Yes