missing translation for 'onlineSavingsMsg'
Learn More

Farnesol, mixture of isomers, 96%

CAS: 4602-84-0 | C15H26O | 222.37 g/mol

Supplier:  Thermo Scientific Chemicals A1931614

Quantity 25 g

 View more versions of this product

Catalog No. AAA1931614


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It can be used to induce apoptosis in cell cultures. It is also used as an antimicrobial agent. It is used in perfumes and as a flavoring agent.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

4602-84-0
222.37
CRDAMVZIKSXKFV-YFVJMOTDSA-N
1549109
(2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
C15H26O
MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
CHEBI:35966
CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Farnesol
4602-84-0
159°C to 160°C (10 mmHg)
Pleasant
1.489
MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
Light sensitive
e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
CRDAMVZIKSXKFV-YFVJMOTDSA-N
(2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
1549109
222.37
mixture of isomers
0.887
146°C (295°F)
C15H26O
25 g
1763926
14,3937
Not miscible or difficult to mix in water.
CC(C)=CCC\C(C)=C\CC\C(C)=C\CO
222.37
CHEBI:35966
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 225-004-1

RTECSNumber : JR4979000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions