accessibility menu, dialog, popup

UserName

CH3CH3CH3CH3OH(E)(E)

CAS RN 4602-84-0

IUPAC Name: (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
Synonyms: (E)-farnesol trans,trans-farnesol trans,trans-α-farnesol
Molecular Weight (g/mol): 222.37
Molecular Formula: C15H26O
InChi Key: CRDAMVZIKSXKFV-YFVJMOTDSA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CO
Boiling Point 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)