missing translation for 'onlineSavingsMsg'
Learn More

FAPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113305 Supplier Thermo Fisher Scientific Supplier No. PA5113305

FAPP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications

Antigen FAPP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PLEKHA8
Gene Accession No. Q80W71, Q96JA3
Gene Alias 6330400G01; AA517676; BC052360; FAPP2; Fapp-2; hFAPP2; MGC3358; PH domain-containing family A member 8; phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing A8; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; pleckstrin homology domain-containing family A member 8; PLEKHA8; RGD1563940; Serologically defined breast cancer antigen NY-BR-86
Gene Symbols PLEKHA8
Host Species Rabbit
Immunogen Recombinant Human Pleckstrin homology domain-containing family A member 8 protein (1-300AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231999, 84725
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less