missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCE Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302125AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
As part of the Fanconi anemia (FA) complex functions in DNA cross-links repair. Required for the nuclear accumulation of FANCC and provides a critical bridge between the FA complex and FANCD2.Spécifications
| FANCE | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9HB96 | |
| FANCE | |
| Between 200 and 250 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| FANCE | |
| FAE; FACE; | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2178 | |
| 4° C | |
| Liquid |