missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FANCA/CEN16q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
$1948.23 - $1948.23
Specifications
| Content And Storage | Store at 4°C in the dark. |
|---|---|
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Format | Tube |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 2175 |
Specifications
| Store at 4°C in the dark. | |
| Tube | |
| 2175 | |
| Human |
| DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
| FISH-Ce | |
| DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
| Liquid |