missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FANCA/CEN16q FISH Probe
Description
- The FANCA/CEN16q FISH Probe is designed for the detection of the human FANCA gene and the centromeric region of chromosome 16 in various sample types.
- Chromosome Region: 16q24.3/16q12.1
- Fluorophore: Texas Red/FITC
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Detection Method | Fluorescence |
| Format | Tube |
| Label or Dye | FITC |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 2175 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Product Type | FISH Probe |
| Quantity | 100 μL |
| Show More |