Learn More
StatLab Fan Pad-GL Formalin Neutralizing Pads
Description
- After absorption, formaldehyde is converted to a non-hazardous polymer eliminating harmful vapors
- Fan Pad minis are also especially useful in the clean up of small formalin spills
Specifications
Specifications
| Material | Polymer |
| Type | Absorbent Pad |
| Disposable | Limited-use |
| Size | Mini |
| Weight Capacity | Heavyweight |