missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Famoxadone Recombinant Mouse Monoclonal Antibody (4E11)

Catalog No. MA566511
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566511 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566511 Supplier Invitrogen™ Supplier No. MA566511

Mouse Recombinant Monoclonal Antibody

Famoxadone Recombinant Monoclonal Antibody for ELISA

Famoxadone is a broad-spectrum fungicide belonging to the oxazolidinone class, utilized primarily in agriculture to protect crops such as grapes, potatoes, and tomatoes from various fungal diseases, including downy mildew and early blight. Structurally, famoxadone is characterized by an oxazolidinone ring, which imparts its ability to inhibit mitochondrial electron transport at the cytochrome bc1 complex, disrupting cellular energy production in target fungi. This mechanism effectively halts fungal growth and sporulation while demonstrating potent residual activity, providing extended protection against infections. Famoxadone is typically applied as a foliar spray, enabling efficient coverage and absorption by plant surfaces. Concerns regarding its environmental persistence and impact on non-target organisms have led to rigorous assessments and regulations governing its use. Research explores optimizing application techniques and resistance management approaches to ensure its sustainable use within integrated crop protection strategies.

Specifications

Antigen Famoxadone
Applications ELISA
Classification Recombinant Monoclonal
Clone 4E11
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Conjugate of FAM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less