missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Famotidine 98.0+%, TCI America™
$267.61 - $668.03
Chemical Identifiers
| CAS | 76824-35-6 |
|---|---|
| Molecular Formula | C8H15N7O2S3 |
| Molecular Weight (g/mol) | 337.435 |
| InChI Key | XUFQPHANEAPEMJ-UHFFFAOYSA-N |
| Synonym | famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin |
| PubChem CID | 5702160 |
| ChEBI | CHEBI:4975 |
| IUPAC Name | 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide |
| SMILES | C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N |
Chemical Identifiers
| 76824-35-6 | |
| 337.435 | |
| famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin | |
| CHEBI:4975 | |
| C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N |
| C8H15N7O2S3 | |
| XUFQPHANEAPEMJ-UHFFFAOYSA-N | |
| 5702160 | |
| 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide |
Specifications
| 76824-35-6 | |
| White-Yellow | |
| 5 g | |
| XUFQPHANEAPEMJ-UHFFFAOYSA-N | |
| 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide | |
| 5702160 | |
| 337.44 | |
| Crystalline Powder |
| 160°C | |
| C8H15N7O2S3 | |
| famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin | |
| C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N | |
| 337.435 | |
| CHEBI:4975 | |
| ≥98.0% (HPLC,T) | |
| Famotidine |
Safety and Handling
RTECSNumber : UA2300000
TSCA : No