missing translation for 'onlineSavingsMsg'
Learn More

Famciclovir Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012363
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012363 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012363 Supplier Merck Emd Millipore Supplier No. PHR16961G
Famciclovir is an antiviral drug, which is predominantly used in the treatment of herpes virus infection. It is an inactive prodrug, which on administration, undergoes rapid biotransformation to penciclovir.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 104227-87-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H19N5O4
MDL Number MFCD00866964
Synonym Famciclovir; 2-(2-(2-Amino-9 H-purin-9-yl)ethyl)-1,3-propanediol diacetate ester; BRL 42810
Type Certified Reference Material
Quantity 1 g
Formula Weight 321.33
Grade Certified Reference Material