missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM50A Recombinant Rabbit Monoclonal Antibody (23GB5830)
GREENER_CHOICE

Catalog No. MA552924
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552924 100 μL
MA552925 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552924 Supplier Invitrogen™ Supplier No. MA552924

Rabbit Recombinant Monoclonal Antibody

FAM50A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

May be a DNA-binding protein or transcriptional factor.

Specifications

Antigen FAM50A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5830
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FAM50A
Gene Accession No. Q14320, Q9WV03
Gene Alias 9F; D0HXS9928E; DXS9928E; FAM50A; family with sequence similarity 50 member A; family with sequence similarity 50, member A; HXC26; HXC-26; Protein FAM50A; Protein HXC-26; Protein XAP-5; RGD1560964; XAP5; XAP-5; XX-FW81657B9.2
Gene Symbols FAM50A
Host Species Rabbit
Immunogen A synthesized peptide derived from human FAM50A (1-45AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108160, 293862, 9130
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less