missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM20A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149927
Description
FAM20A Polyclonal Antibody for Western Blot, ICC/IF, IHC
This locus encodes a protein that is likely secreted and may function in hematopoiesis. A mutation at this locus has been associated with amelogenesis imperfecta and gingival hyperplasia syndrome. Alternatively spliced transcript variants have been identified.Chemical Identifiers
| FAM20A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8CID3, Q96MK3 | |
| 208659, 303635, 54757 | |
| Recombinant Family With Sequence Similarity 20, Member A (FAM20A) (Lys368-Val523). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FAM20A | |
| AI1G; AI606893; AIGFS; FAM20A; family with sequence similarity 20 member A; family with sequence similarity 20, member A; FP2747; ortholog of human family with sequence similarity 20, member A FAM20A; protein FAM20A; pseudokinase FAM20A; RGD1306364; UNQ9388/PRO34279 | |
| FAM20A | |
| Primary | |
| Protein A |
Specifications
| FAM20A | |
| Polyclonal | |
| Unconjugated | |
| FAM20A | |
| AI1G; AI606893; AIGFS; FAM20A; family with sequence similarity 20 member A; family with sequence similarity 20, member A; FP2747; ortholog of human family with sequence similarity 20, member A FAM20A; protein FAM20A; pseudokinase FAM20A; RGD1306364; UNQ9388/PRO34279 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 208659, 303635, 54757 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8CID3, Q96MK3 | |
| FAM20A | |
| Recombinant Family With Sequence Similarity 20, Member A (FAM20A) (Lys368-Val523). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |