missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM198B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552406
Description
Immunogen sequence: PRTRRNLLLG TACAIYLGFL VSQVGRASLQ HGQAAEKGPH RSRDTAEPSF PEIPLDGTLA PPESQGNGST LQPNVVYITL RSKRSKPANI RGTVKPKRRK KHAVASAAPG QEALVGPSLQ PQEAAREADA VAPGYA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
FAM198B is a protein coding gene.Chemical Identifiers
| FAM198B | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| FAM198B | |
| 1110032E23Rik; 2210419I08Rik; AD021; AD036; AV011458; C4orf18; Ened; Expressed in nerve and epithelium during development; FAM198B; family with sequence similarity 198 member B; family with sequence similarity 198, member B; gask1b; GaskbB; golgi associated kinase 1B; Golgi-associated kinase 1B; LOW QUALITY PROTEIN: protein FAM198B; protein ENED; protein ENED; protein FAM198B; protein FAM198B; UNQ2512/PRO6001 | |
| FAM198B | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWH4 | |
| 51313 | |
| Recombinant protein corresponding to Human FAM198B. Recombinant protein control fragment (Product #RP-89728). | |
| Antibody |
Specifications
| FAM198B | |
| Polyclonal | |
| Unconjugated | |
| FAM198B | |
| 1110032E23Rik; 2210419I08Rik; AD021; AD036; AV011458; C4orf18; Ened; Expressed in nerve and epithelium during development; FAM198B; family with sequence similarity 198 member B; family with sequence similarity 198, member B; gask1b; GaskbB; golgi associated kinase 1B; Golgi-associated kinase 1B; LOW QUALITY PROTEIN: protein FAM198B; protein ENED; protein ENED; protein FAM198B; protein FAM198B; UNQ2512/PRO6001 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51313 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWH4 | |
| FAM198B | |
| Recombinant protein corresponding to Human FAM198B. Recombinant protein control fragment (Product #RP-89728). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |