Learn More
Falcon™ High Density Permeable Support for 24-well Plate
With 0.4μm Translucent high density PET membrane
Supplier:  Falcon™ 353495
Catalog No. 08-771-17
Only null left
Description
Description
  • Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12, and 24 well
  • A broad selection of membrane pore sizes, 0.4, 1.0, 3.0, and 8.0 μm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Specifications
Specifications
Flat
PET
Sterile
Multi-well Cell Culture Plate
48/Cs.
0.4 μm
0.3 cm2
Round
Cell Culture
Transport, diffusion and secretion of small molecules into, out of, or through a cell monolayer. Barrier function (Transepithelial Electrical Resistance (TEER) measurements). Drug bioavailability
1/Pk.
6.4 mm (Membrane)
Product Suggestions
Product Suggestions