missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Feeder Tray for FluoroBlok™ HTS 24 Well Multiwell plates
GREENER_CHOICE

Catalog No. 08781
Change view
Click to view available options
Well Count:
24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
08781 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08781 Supplier Falcon™ Supplier No. 351186
Only null left

Polystyrene cell media tray for feeding cell-based assays.

Polystyrene cell media tray for feeding cell-based assays

Storage: Store at room temperature, away from direct sunlight.

Specifications

Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Non-treated
Type Feeder Tray
Well Count 24
For Use With (Equipment) FluoroBlok™ HTS 24 Well Multiwell plates, Falcon™ 24 Well Multiwell plates
Includes Lid
No. per Case 5/Cs.
No. per Pack 5/Pk.
Pore Size Not applicable
Quantity 5/Pk.
Storage Requirements Store at room temperature, away from direct sunlight
Supportive Membrane Polyethylene Terephthalate
Show More Show Less