missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor XIa Native Protein

Catalog No. PIRP43122
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43122 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43122 Supplier Invitrogen™ Supplier No. RP43122

Native Protein

RP-43122 contains native human Factor XIa protein. RP-43122 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!

Specifications

Accession Number P03951
Concentration 3.1 mg/mL
For Use With (Application) Control
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 2160
Molecular Weight (g/mol) 160 kDa
Name Human Factor XIa
Quantity 50 μg
Source Human
Regulatory Status RUO
Gene Alias coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; F11a; Factor 11; Factor11; FXI; Plasma thromboplastin antecedent; PTA
Common Name Factor XIa
Gene Symbol F11
Biological Activity Specific Activity = 636 U/mg; determined by Factor XI clotting assay where 1 U equals the Factor XI activity in 1 mL of normal human plasma.
Conjugate Unconjugated
Species Human
Content And Storage -20°C
Form Liquid
Protein Subtype Native
Show More Show Less