missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor XI Native Protein

Numéro de catalogue. PIRP43075
Modifier l'affichage
Click to view available options
Quantité:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIRP43075 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIRP43075 Fournisseur Invitrogen™ Code fournisseur RP43075

Native Protein

RP-43075 contains native human Factor XI protein. RP-43075 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!

This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality.

Spécifications

Numéro d'enregistrement P03951
Concentration 7.0 mg/mL
À utiliser avec (application) Control
Formule 50% water with 50% glycerol and no preservative
ID du gène (Entrez) 2160
Poids moléculaire (g/mol) 160 kDa
Nom Human Factor XI
Quantité 50 μg
Sources Human
Statut réglementaire RUO
Alias du gène 1600027G01Rik; AI503996; Cf11; Coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; FXI; LRRGT00086; plasma thromboplastin antecedent; PTA
Nom usuel Factor XI
Symbole de gène F11
Activité biologique Specific Activity =372 U/mg; determined by Factor XI clotting assay where 1 U equals the Factor XI activity in 1 mL of normal human plasma.
Conjugué Unconjugated
Espèces Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Forme Liquid
Sous-type de protéine Native
Afficher plus de résultats Afficher moins de résultats