missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor X Native Protein

Catalog No. PIRP43073
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43073 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43073 Supplier Invitrogen™ Supplier No. RP43073

Native Protein

RP-43073 contains native bovine Factor X protein. RP-43073 may be used as a positive control in many immunological assays.

F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.

Specifications

Accession Number P00743
Concentration 6.0 mg/mL
For Use With (Application) Control
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 280787
Molecular Weight (g/mol) 55.1 kDa
Name Bovine Factor X
Quantity 100 μg
Source Bovine
Regulatory Status RUO
Gene Alias Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor x protein; F10; Factor 10; Factor x heavy chain; Factor x light chain; factor Xa; Factor10; FX; FXA; prothrombinase; RP11-98F14.9; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product
Common Name Factor X
Gene Symbol F10
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less