missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Factor IX Native Protein
Description
RP-43069 contains native human Factor IX protein. RP-43069 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifications
Specifications
| Accession Number | P00740 |
| Concentration | 6.0 mg/mL |
| For Use With (Application) | Control |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene ID (Entrez) | 2158 |
| Molecular Weight (g/mol) | 55 kDa |
| Name | Human Factor IX |
| Quantity | 100 μg |
| Source | Human |
| Gene Alias | AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8 |
| Show More |