Learn More
Invitrogen™ Human FABP-3 ELISA Kit
ELISA
$896.00 - $6640.00
Specifications
| Accession Number | P05413 |
|---|---|
| Assay Range | 156.3 to 5000 pg/mL |
| Assay Sensitivity | 26.88 pg/ml |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BMS2263
|
Invitrogen™
BMS2263 |
96 Tests |
Each for $896.00
|
|
|||||
BMS2263TEN
|
Invitrogen™
BMS2263TEN |
10 x 96 Tests |
Each for $6,640.00
|
|
|||||
Description
The Human FABP-3 ELISA quantitates Hu FABP-3 in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP-3.
Principle of the method
The Human FABP-3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Fatty acid binding proteins (FABP) are small (approximately 13-14 kDa) intracellular proteins with a high degree of tissue specificity. FABPs are a class of cytoplasmic proteins that bind long chain fatty acids. They are abundantly present in various cell types and seem to play an important role in the intracellular utilization of fatty acids. There are at least six distinct types of FABP, each showing a specific pattern of tissue expression. FABP leaks, due to its small size, rapidly out of is chemically damaged dying cells leading to a rise in serum levels. Liver FABP is a sensitive marker for cell damage of liver cells in vitro and in vivo. Is chemically damaged tissues are characterized histologically by absence (or low presence) of FABP facilitating recognition of such areas. Next to this L FABP is a marker for rapid hepatocyte lysis in vitro (as for example in toxicology assays) and for detection of liver damage during and after transplantation.Specifications
| P05413 | |
| 26.88 pg/ml | |
| ELISA | |
| Human | |
| 2170 | |
| 6.2% | |
| Pre-coated 96 well plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 156.3 to 5000 pg/mL | |
| HRP | |
| Cell Lysates, Plasma, Serum, Supernatant | |
| Colorimetric Microplate Reader | |
| FABP3 | |
| 3.9 | |
| HRP | |
| Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FABP3 | |
| 4 hrs |