missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FA156 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5112242

Catalog No. PIPA5112242


Only null left

Description

Description

Specificity: Human FA156.

Specifications

Chemical Identifiers

FA156
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
FAM156A
FAM156A; FAM156B; family with sequence similarity 156 member A; family with sequence similarity 156, member A; PP12994; PRO0659; protein FAM156A; protein FAM156A/FAM156B; TMEM29; TMEM29B; transmembrane protein 29; transmembrane protein 29/29B
FAM156A
Primary
Antigen affinity chromatography, Protein A
1 mg/mL
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.03% ProClin 300; pH 7
Q8NDB6
29057
E. coli-derived Human FAM156A fragment.
Antibody

Specifications

FA156
Polyclonal
Unconjugated
FAM156A
FAM156A; FAM156B; family with sequence similarity 156 member A; family with sequence similarity 156, member A; PP12994; PRO0659; protein FAM156A; protein FAM156A/FAM156B; TMEM29; TMEM29B; transmembrane protein 29; transmembrane protein 29/29B
Rabbit
Antigen affinity chromatography, Protein A
RUO
29057
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.03% ProClin 300; pH 7
Q8NDB6
FAM156A
E. coli-derived Human FAM156A fragment.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions