Learn More
Epredia™ Laser Cassette Printer Accessories
Designed for use with Epredia™ Laser Cassette Printers, these accessories will help support all your printing needs.
$668.96 - $20,509.16
Specifications
Description Laser Cassette Printer Accessories
View More Specs
Products 9
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
8000042
View Documents Promotion Details
Epredia™
8000042
VEGA Laser Printer VEGA Sorter V2 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
8000043
View Documents Promotion Details
Epredia™
8000043
VEGA Laser Printer VEGA DUO V2 System N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
9000704
View Documents Promotion Details
Epredia™
9000704
VEGA Laser Printer DUO Tray (Wood)
Each for $668.96
Only null left
9000784
View Documents Promotion Details
Epredia™
9000784
NOVA Laser Printer NOVA Additional Hopper
Each for $5,541.04
Only null left
9001140
View Documents Promotion Details
Epredia™
9001140
VEGA Laser Printer VEGA Side Delivery System
Each for $3,515.85
Only null left
9001150
View Documents Promotion Details
Epredia™
9001150
NOVA Laser Printer NOVA Side Delivery System
Each for $3,515.85
Only null left
9001327
View Documents Promotion Details
Epredia™
9001327
VEGA Laser Printer Hopper Carousel
Each for $8,767.24
Only null left
9001387
View Documents Promotion Details
Epredia™
9001387
VEGA Laser Printer Sorter Sorter Collector Tray
Each for $4,452.25
Only null left
9001547
View Documents Promotion Details
Epredia™
9001547
VEGA Laser Printer DUO Unit DUO Tray (Metal)
Case of 4 Each for $2,171.11
Only null left
Description
Description
VEGA™ Sorter:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Organizes batch printing mode for busy mid to large laboratories.
  • Achieve prioritized printing as needed outside of the print queue for urgent patient samples.
  • Positioned at the base of the VEGA™ printer, keeps the footprint small saving bench space.
  • Permits up to 25 cassettes to be pre-printed and sequentially released into the collector tray.
Collector:
  • Collection tray for use with Vega Cassette Sorter to keep cassettes in an organized manner.
  • Cassettes collected in a vertical fashion for easy visual recognition.
VEGA™ Duo Unit:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Partnered with the Vega printer, permits up to 2 users to print jointly in busy laboratories while in both batch and print-on-demand modes keeping printed cassettes organized by user.
  • Up to 18 cassettes can be pre-printed and sequentially released into the collection trays keeping up with workflow demand.
  • Saves laboratories the need for additional printers lowing capital expense.
Hopper:
  • Holds up to 80 additional cassettes for easy exchange to a different color or style of cassettes when using the Nova Laser Printer.
Carousel:
  • Utilize six additional hoppers, holding up to a total of 480 cassettes, to be partnered with the Vega Laser Printer.
  • Permits for easy replenishment or exchange of cassettes for continual workflow without interruptions for busy laboratories.
Side Delivery System:
  • Available for both VEGA™ and NOVA printers.
  • Keeps printed cassettes in an organized fashion when printing in either batch or print-on-demand modes.
  • Delivers cassettes directly onto collection trays to eliminate workflow delays.
Trays:
  • Metal – two trays each holding up to 10 cassettes paired with the Vega Duo Unit to keep cassettes in an organized manner.
  • Wooden trays more environmentally friendly.
  • Two trays each holding up to 10 cassettes.
Specifications
Specifications
Laser Cassette Printer Accessories