Learn More
Epredia™ Laser Cassette Printer Accessories
Catalog No. 9001547
Change view
Click to view available options
For Use With (Equipment):
NOVA Laser Printer
VEGA Laser Printer
VEGA Laser Printer DUO Unit
VEGA Laser Printer Sorter
Product Type:
DUO Tray (Metal)
DUO Tray (Wood)
Hopper Carousel
NOVA Additional Hopper
NOVA Side Delivery System
Sorter Collector Tray
VEGA DUO V2 System
VEGA Side Delivery System
VEGA Sorter V2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
9001547 VEGA Laser Printer DUO Unit DUO Tray (Metal)
8000042 VEGA Laser Printer VEGA Sorter V2
8000043 VEGA Laser Printer VEGA DUO V2 System
9000704 VEGA Laser Printer DUO Tray (Wood)
9000784 NOVA Laser Printer NOVA Additional Hopper
9001140 VEGA Laser Printer VEGA Side Delivery System
9001150 NOVA Laser Printer NOVA Side Delivery System
9001327 VEGA Laser Printer Hopper Carousel
9001387 VEGA Laser Printer Sorter Sorter Collector Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 9001547 Supplier Epredia™ Supplier No. 9001547
Designed for use with Epredia™ Laser Cassette Printers, these accessories will help support all your printing needs.
VEGA™ Sorter:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Organizes batch printing mode for busy mid to large laboratories.
  • Achieve prioritized printing as needed outside of the print queue for urgent patient samples.
  • Positioned at the base of the VEGA™ printer, keeps the footprint small saving bench space.
  • Permits up to 25 cassettes to be pre-printed and sequentially released into the collector tray.
Collector:
  • Collection tray for use with Vega Cassette Sorter to keep cassettes in an organized manner.
  • Cassettes collected in a vertical fashion for easy visual recognition.
VEGA™ Duo Unit:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Partnered with the Vega printer, permits up to 2 users to print jointly in busy laboratories while in both batch and print-on-demand modes keeping printed cassettes organized by user.
  • Up to 18 cassettes can be pre-printed and sequentially released into the collection trays keeping up with workflow demand.
  • Saves laboratories the need for additional printers lowing capital expense.
Hopper:
  • Holds up to 80 additional cassettes for easy exchange to a different color or style of cassettes when using the Nova Laser Printer.
Carousel:
  • Utilize six additional hoppers, holding up to a total of 480 cassettes, to be partnered with the Vega Laser Printer.
  • Permits for easy replenishment or exchange of cassettes for continual workflow without interruptions for busy laboratories.
Side Delivery System:
  • Available for both VEGA™ and NOVA printers.
  • Keeps printed cassettes in an organized fashion when printing in either batch or print-on-demand modes.
  • Delivers cassettes directly onto collection trays to eliminate workflow delays.
Trays:
  • Metal – two trays each holding up to 10 cassettes paired with the Vega Duo Unit to keep cassettes in an organized manner.
  • Wooden trays more environmentally friendly.
  • Two trays each holding up to 10 cassettes.
Specifications
Product Type DUO Tray (Metal)
Description Laser Cassette Printer Accessories
For Use With (Equipment) VEGA Laser Printer DUO Unit
Material Metal