missing translation for 'onlineSavingsMsg'
Learn More

F4/80 Monoclonal Antibody (BM8), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$624.38 - $1322.02

Specifications

Antigen F4/80
Clone BM8
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
40-748-0182
View Documents
Invitrogen™
407480182
100 μg
Each for $1,322.02
Only null left
 
40-748-0180
View Documents
Invitrogen™
407480180
25 μg
Each for $624.38
Only null left
 
Description

Description

F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

F4/80
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3
IgG2a κ
Affinity chromatography
Antibody
BM8
Flow Cytometry
Brilliant Violet 711
Rat
Mouse
Q61549
13733
Primary
4°C, store in dark, DO NOT FREEZE!
ADGRE1
Videos
SDS
Documents

Documents