missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ F1 OIML Class Standard Weight Set (F1 AL)

Catalog No. 01912676
Change view
Click to view available options
Weight (Metric):
1 mg to 2 kg
1 mg to 5 kg
1 to 50 g
1 to 500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
01912676 1 to 500 g
01912673 1 mg to 2 kg
01912674 1 mg to 5 kg
01912675 1 to 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01912676 Supplier Mettler Toledo™ Supplier No. 158886

F1 OIML class weight set includes accessories for handling and cleaning, and is housed in an aluminum box.

  • Without calibration certificate
  • For standard environments: High quality weights for regular routine testing without the need for traceability
  • High-grade stainless steel: Low magnetic permeability and high corrosion resistance ensure reliable values

Specifications

Product Type Test Weight
Material Stainless Steel
Case Material Aluminum
Weight (Metric) 1 to 500 g
Density 8000 (± 60) kg/m3
Includes 12 Pieces; Without certificate