missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes: PROMO

Catalog No. 14488038TI
Change view
Click to view available options
Volume (Metric):
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
8
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
14488038TI 30 to 300 μL 12
14488032TI 5 to 50 μL 8
14488036TI 5 to 50 μL 12
14488033TI 10 to 100 μL 8
14488034TI 30 to 300 μL 8
14488037TI 10 to 100 μL 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14488038TI Supplier Thermo Scientific™ Supplier No. P4661180
Get Up to 65% Off Pipettes
View all promotions

Innovative locking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.

Features

  • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip enhanced accuracy and precision
  • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette

Ergonomic Design

  • Offering low attachment, ejection and plunger forces for safe and comfortable use

Remarkably Lightweight

  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)

Adjustable Comfort

  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles

Super Blow-out

  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action

Soft-touch Tip Ejection

  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle pus

Volume Lock Adjustment

  • The secure volume setting prevents undesirable volume drift during operation

Industry-leading Lifetime Warranty

  • With convenient online registration

Specifications

Compatible Tips F1-ClipTip 300
Color Orange
For Use With F1-ClipTip™ 300
Model F1-ClipTip™
Number of Channels 12
Product Line F1-ClipTip
Type Pipette
Volume (Metric) 30 to 300 μL
Increments 1 μL