Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes: PROMO
Innovative locking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Supplier:  Thermo Scientific™ P4661160
 View more versions of this product
Catalog No. 14488036TI
Only null left
Explore available promotions
Description
Description
Features
  • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip enhanced accuracy and precision
  • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette
  • Ergonomic Design
  • Offering low attachment, ejection and plunger forces for safe and comfortable use
  • Remarkably Lightweight
  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
  • Adjustable Comfort
  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
  • Super Blow-out
  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
  • Soft-touch Tip Ejection
  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle pus
  • Volume Lock Adjustment
  • The secure volume setting prevents undesirable volume drift during operation
  • Industry-leading Lifetime Warranty
  • With convenient online registration
  • Specifications
    Specifications
    1.5 to 5%
    50
    12-channel
    F1-ClipTip™ 50
    12
    Pipette
    0.1 μL
    The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    F1-ClipTip 50
    Violet
    F1-ClipTip™
    F1-ClipTip
    5 to 50 μL
    0.7 to 2%
    Product Suggestions
    Product Suggestions