Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes: PROMO
Innovative locking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Supplier: Thermo Scientific™ P4661140
Description
FeaturesErgonomic Design
Remarkably Lightweight
Adjustable Comfort
Super Blow-out
Soft-touch Tip Ejection
Volume Lock Adjustment
Industry-leading Lifetime Warranty
Specifications
| F1-ClipTip 300 | |
| F1-ClipTip™ 300 | |
| 8 | |
| Pipette | |
| 1 μL |
| Orange | |
| F1-ClipTip™ | |
| F1-ClipTip | |
| 30 to 300 μL |