Learn More
Invitrogen™ F(ab)-Rabbit anti-Goat IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 488
Description
F(ab)-Rabbit anti-Goat IgG (H+L) Superclonal™ Recombinant Secondary Antibody detects both the Fab2 and Fc fragments of Goat IgG. It showed no cross-reactivity to various species immunoglobulins tested. This Fab fragment antibody contains a hexa-histidine tag, is expressed in vitro and purified as a recombinant construct. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. F(ab) fragments offer several advantages over full-length IgGs due to their smaller molecular size (~50 kDa vs. ~150 kDa), such as improved tissue penetration. The absence of the Fc region reduces nonspecific binding and background staining in Fc receptor-expressing cells and tissues, while also enabling increased flexibility in multiplexing assays. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Spécifications
Spécifications
| Antigène | Goat IgG (H+L) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Superclonal |
| Concentration | 1 mg/mL |
| Conjugué | Alexa Fluor Plus 488 |
| Formule | Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3 |
| Espèces hôtes | Rabbit |
| Immunogène | Purified Goat IgG. |
| Méthode de purification | Affinity chromatography |
| Quantité | 250 μL |
| Afficher plus de résultats |