missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Rabbit anti-Rat IgG (H+L) Secondary Antibody, AP
Description
PA1-29928 detects Rat IgG (H+L) from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Rat IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | TBS with 1mM MgCl2, 0.1mM ZnCl2, 50% glycerol, 1% BSA and 0.05% sodium azide; pH 8 |
| Host Species | Rabbit |
| Immunogen | Rat IgG whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 250 μg |
| Show More |