missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, PE

Catalog No. A10543
Encompass
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10543 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10543 Supplier Invitrogen™ Supplier No. A10543

F(ab')2-Goat Polyclonal Secondary Antibody

F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody for Flow

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate PE
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Goat
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Purified
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less