missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Goat anti-Human IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, FITC

Catalog No. A24489
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24489 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24489 Supplier Invitrogen™ Supplier No. A24489

F(ab')2-Goat Polyclonal Secondary Antibody

F(ab')2-Goat anti-Human IgM (Heavy chain) Cross-Adsorbed Secondary Antibody for Flow, ICC/IF

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgM (Heavy chain) Cross-Adsorbed
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.05% sodium azide; pH 7.8
Host Species Goat
Immunogen Human Mu immunoglobin.
Purification Method Affinity Chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less