missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Syrian/Armenian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody
Description
F(ab')2 fragments were prepared by pepsin digestion. The antibody has been cross adsorbed against both mouse and rat immunoglobulins to minimize cross-reactivity.
Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Syrian/Armenian Hamster IgG (H+L) Cross-Adsorbed |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Host Species | Goat |
| Immunogen | Hamster IgG. |
| Purification Method | Affinity chromatography |
| Quantity | 400 μg |
| Show More |