missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Donkey anti-Rabbit IgG (H+L) Secondary Antibody, PE, eBioscience™

Catalog No. 501128861
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128861 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501128861 Supplier Invitrogen™ Supplier No. 12473981
Only null left

F(ab')2-Donkey Polyclonal Secondary Antibody

The anti-rabbit IgG polyclonal antibody reacts with the heavy and light chains of rabbit IgG, with minimal cross reaction to bovine, guinea pig, Syrian hamster, horse, human, mouse, rat and sheep serum proteins. Reactions to non-immunoglobulin rabbit serum proteins have not been detected. Applications Reported: This polyclonal antibody has been reported for use in flow cytometric analysis. Applications Tested: This polyclonal antibody has been tested by flow cytometric analysis to detect polyclonal rabbit antibodies. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (H+L)
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate PE
Formulation PBS with BSA and 0.05% sodium azide; pH 7.6
Host Species Donkey
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype Ig
Show More Show Less