missing translation for 'onlineSavingsMsg'
Learn More

F.A.M.E. Mix, C4-C24, Certified Reference Material, wt. (varied), MilliporeSigma™ Supelco™

Catalog No. 111020809
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020809 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020809 Supplier Merck Emd Millipore Supplier No. 189191AMP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complementary products:SyringesVialsGas Purification and Gas ManagementInlet Liners, Glass Wool, and Specialized Hand ToolsSepta and Specialized Hand ToolsColumn Ferrules, Nuts, and Specialized Hand Tools

Specifications

Physical Form Solid
Quantity 100 mg
Grade Certified Reference Material
Concentration or Composition (by Analyte or Components) cis-13,16-Docosadienoic acid methyl ester 2 wt. %; cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 2 wt. %; cis-11,14-Eicosadienoic acid methyl ester 2 wt. %; cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester 2 wt. %; cis-8,11,14-Eicosatrienoic acid methyl ester 2 wt. %; cis-11,14,17-Eicosatrienoic acid methyl ester 2 wt. %; Methyl arachidate 4 wt. %; Methyl arachidonate 2 wt. %; Methyl behenate 4 wt. %; Methyl butyrate 4 wt. %; Methyl decanoate 4 wt. %; Methyl cis-13-docosenoate 2 wt. %; Methyl dodecanoate 4 wt. %; Methyl cis-11-eicosenoate 2 wt. %; Methyl elaidate 2 wt. %; Methyl heneicosanoate 2 wt. %; Methyl heptadecanoate 2 wt. %; Methyl cis-10-heptadecenoate 2 wt. %; Methyl hexanoate 4 wt. %; Methyl linoleate 2 wt. %; Methyl linolelaidate 2 wt. %; Methyl linolenate 2 wt. %; Methyl gamma-linolenate 2 wt. %; Methyl myristate 4 wt. %; Methyl myristoleate 2 wt. %; Methyl octanoate 4 wt. %; Methyl oleate 4 wt. %; Methyl palmitate 6 wt. %; Methyl palmitoleate 2 wt. %; Methyl pentadecanoate 2 wt. %; Methyl cis-10-pentadecenoate 2 wt. %; Methyl stearate 4 wt. %; Methyl tetracosanoate 4 wt. %; Methyl cis-15-tetracosenoate 2 wt. %; Methyl tricosanoate 2 wt. %; Methyl tridecanoate 2 wt. %; Methyl undecanoate 2 wt. %