missing translation for 'onlineSavingsMsg'
Learn More

Innovatek EZTest Steam G. stearothermophilus

Innovatek EZTest - Steam G.stearothermophilus is an easy-to-use test for monitoring sterilization

$378.11 - $1,270.29

Specifications

Color Yellow Indicator
For Use With (Application) Monitoring Sterilization
Model EZTest™
Temperature (Metric) 57°C
Heating Time 24 hours at 57°C
View More Specs

Products 4
Catalog Number Mfr. No. Population Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Population Quantity Price Quantity & Availability  
29801075
View Documents Promotion Details
Innovatek
4140EZS/525
105 25 Vials/Box
Pack of 25 Each for $378.11
Only null left
Edge
 
29801074
View Documents Promotion Details
Innovatek
4140EZS/5
105 100 Vials/Box
Pack of 100 Each for $702.24
Only null left
 
29801077
View Documents Promotion Details
Innovatek
4140 EZS 625
106 25 Vials/Box
Pack of 25 Each for $638.29
Only null left
 
29801076
View Documents Promotion Details
Innovatek
4140 EZS 6
106 100 Vials/Box
Pack of 100 Each for $1,270.29
Only null left
 
Description

Description

  • EZTest is a self-contained biological indicator (BI) for monitoring sterilization
  • Bacterial spores on a carrier material packaged within a small, thermoplastic culture tube
  • Sealed-glass ampoule of specially formulated soybean casein digest culture medium
    • Color indicator turns noticeably yellow when growth occurs
  • Easy-to-use in-office; no laboratory transfer needed
  • Dramatic visual results within 24 hours (48 hours for ethylene oxide)
Specifications

Specifications

Yellow Indicator
EZTest™
24 hours at 57°C
Monitoring Sterilization
57°C
Soybean Casein Digest Culture Medium, Color Indicator
Videos
SDS
Documents

Documents