missing translation for 'onlineSavingsMsg'
Learn More

Innovatek EZTest Steam G. stearothermophilus

Catalog No. 29801074
Change view
Click to view available options
Population:
105
106
Quantity:
100 Vials/Box
25 Vials/Box
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Population Quantity
29801074 105 100 Vials/Box
29801075 105 25 Vials/Box
29801077 106 25 Vials/Box
29801076 106 100 Vials/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29801074 Supplier Innovatek Supplier No. 4140EZS/5
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek EZTest - Steam G.stearothermophilus is an easy-to-use test for monitoring sterilization

  • EZTest is a self-contained biological indicator (BI) for monitoring sterilization
  • Bacterial spores on a carrier material packaged within a small, thermoplastic culture tube
  • Sealed-glass ampoule of specially formulated soybean casein digest culture medium
    • Color indicator turns noticeably yellow when growth occurs
  • Easy-to-use in-office; no laboratory transfer needed
  • Dramatic visual results within 24 hours (48 hours for ethylene oxide)

Specifications

Color Yellow Indicator
For Use With (Application) Monitoring Sterilization
Model EZTest™
Temperature (Metric) 57°C
Heating Time 24 hours at 57°C
Includes Soybean Casein Digest Culture Medium, Color Indicator
Population 105
Quantity 100 Vials/Box