Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Assembly, Fritted Disc (Porosity C)
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$1103.13 - $1271.97
Specifications
| Column Type | Flash Chromatography |
|---|---|
| For Use With (Application) | Flash Chromatography |
| Product Line | EZSafe |
Description
EZSafe™ columns are improved low-pressure columns (max. pressure 7 psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy. Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE. GL-45 screw-thread connections ensure positive reliable connections. Eliminates frozen PTFE and glass plugs. Eliminates grease. Choose between standard columns or columns with a fritted disc. The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.Specifications
| Flash Chromatography | |
| Flash Chromatography | |
| EZSafe |
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.