Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Assembly, Fritted Disc (Porosity C)
Catalog No. 111007686
Change view
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
111007686 1000 mL
111007549 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111007686 Supplier MilliporeSigma™ Supelco™ Supplier No. Z4164281EA
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
EZSafe™ columns are improved low-pressure columns (max. pressure 7 psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy. Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE. GL-45 screw-thread connections ensure positive reliable connections. Eliminates frozen PTFE and glass plugs. Eliminates grease. Choose between standard columns or columns with a fritted disc. The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Capacity (Metric) 1000 mL
Product Line EZSafe
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.