missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ EZFluor™ UV 1-step Fluorescent Protein Gel Stain
Fluorescent protein gel stain that requires only a single 5 to 30 minute staining step without fixation.
$390.00 - $1,170.00
Specifications
| Description | Absorbance: 288 nm; Emission: 603 nm |
|---|---|
| Detection | Fluorescence |
| Form | Liquid |
| Storage Requirements | Store at 2 to 8°C, protect from light |
Description
- Ready-to-use luminescent protein gel stain designed for imaging using a UV transilluminator
- Requires only a single 5 to 30 minute staining step without fixation
- Destaining is optional
- Offers safer handling and ease of disposal, because it is an aqueous-based solution that does not contain hazardous methanol or acetic acid
- Passed environmental toxicity testing and is classified as non-hazardous to the environment under CCR Title 22 regulations
- Can detect as little as 1 to 10 ng of protein per band depending on the staining method used
- The results obtained with EZFluor UV protein stain are comparable to those with Oriole™ Fluorescent Gel Stain; however, the EZFluor UV protein stain has a more convenient protocol, does not contain hazardous solvents, and does not cause gel shrinkage
- Staining is fully compatible with mass spectrometry and Edman-based sequencing
- Absorbance: 288 nm
- Emission: 603 nm
Specifications
| Absorbance: 288 nm; Emission: 603 nm | |
| Fluorescence | |
| Liquid | |
| Store at 2 to 8°C, protect from light |