missing translation for 'onlineSavingsMsg'
Learn More

EZ Mix Primary Chlorine Standard Kit, Lovibond™

Catalog No. 15398553
Catalog No. 15398553 Supplier Tintometer Inc Supplier No. 48105510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

For Use With (Equipment) Chlorine Analysis Equipment
Chemical Name or Material EZ Mix Primary Chlorine Standard Kit
ShelfLife One year