missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin

Catalog No. PIA39257
Change view
Click to view available options
Quantity:
100 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA39257 10 x 1 mg
PI21335 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA39257 Supplier Thermo Scientific™ Supplier No. A39257

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

  • Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
  • Medium length - spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass

Spécifications

Product Type Sulfo-NHS-LC-Biotin
Quantity 10 x 1 mg
Content And Storage Store desiccated at -20°C.
Labeling Method Chemical Labeling
Shipping Condition Qualified for Ambient or Wet Ice
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Mid-length
Afficher plus de résultats Afficher moins de résultats
Can I dissolve the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257, 21335) in water for storage?

No, the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257) should not be dissolved in water for storage. We recommended storing EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format at -20 degrees C with desiccant as it is very moisture sensitive. Furthermore, to avoid moisture condensation onto the product, equilibrate vial to room temperature before opening.

Can you provide the shelf-life for EZ-Link Sulfo-NHS-LC-Biotin?

EZ-Link Sulfo-NHS-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Can you provide the shelf life for EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format?

EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (desiccated, at -20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

How does a product with "No-Weigh" format reduce my concern over reagent stability?

The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html

For Research Use Only. Not for use in diagnostic procedures.