missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Micro Sulfo-NHS-Biotinylation Kit

Catalog No. PI21925
Change view
Click to view available options
Quantity:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21925 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21925 Supplier Thermo Scientific™ Supplier No. LSG21925

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.

Highlights:

  • Shorter spacer arm than Sulfo-NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate, or phosphate buffers
  • Negative charge of -SO3 keeps biotinylation localized at the cell surface so reagent does not pass through cell membrane
  • Water solubility eliminates the need for harmful organic solvents
U.S. Patents 5,892,628 and 5,872,261.

Specifications

Description EZ-Link Micro Sulfo-NHS-Biotinylation Kit
Product Type Micro Sulfo-NHS-Biotinylation Kit
Quantity 8 reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Labeling Scale 100 μg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Spacer Pegylated
Labeling Target Antibodies, Proteins
Which EZ-Link Micro Biotinylation Kit is best for my application?

All four Micro Biotinylation Kits will label proteins for use in many applications. Consider the following guidelines for choosing among the kits:

• Choose the Sulfo-NHS-Biotinylation Kit (Cat. No. 21925) when it is important to minimize the molecular mass contributed by the label.
• Choose the Sulfo-NHS-SS-Biotinylation Kit (Cat. No. 21945) only when the downstream application require cleaving the biotin tag with a reducing agent.
• Choose the NHS-PEG4-Biotinylation Kit (Cat. No. 21955) to maximize protein solubility and prevent protein aggregation during storage.
• Choose the Sulfo-NHS-LC-Biotinylation Kit (Cat. No. 21935) for routine purposes when a feature of one of the other reagents is not required. The longer spacer arm offers the possibility of optimizing detection experiments where steric hindrance of biotin binding is an important factor.

How long does it take to biotinylate a protein using the EZ-Link Micro Biotinylation kits?

The entire procedure can be completed in less than 3 hours.

What types of proteins can be labeled with the EZ-Link Micro Biotinylation kits?

Any molecule containing a primary amine can be labeled. Any protein that has been labeled using the standard biotinylation kits, including antibodies and peptides, also can be labeled with the micro kits.

Are the biotinylation reagents in the EZ-Link Micro Biotinylation Kits cleavable?

Only the sulfo-NHS-SS-Biotin is cleavable because it has a disulfide bond in its spacer arm, enabling labeled proteins to be cleaved from the biotin group by treatment with reducing agents.

What are the reaction chemistries of the reagents in the EZ-Link Micro Biotinylation Kits?

The reagents in the Micro kits are amine reactive. NHS esters react with a-amines at the N-termini and the epsilon-amines of lysine side chains at pH 7-9 to form stable amide bonds and the release of NHS.

Are the included biotinylation reagents in the EZ-Link Micro Biotinylation Kits water soluble?

Yes, the four Micro kits contain water-soluble biotinylation reagents; three kits contain sulfo-NHS reagents and one kit contains a polyethylene glycol (PEG) reagent. The sulfo-NHS ester imparts water solubility and is the reaction leaving group. The hydrophilic PEG spacer arm imparts water solubility that is maintained after conjugation.

How many EZ-Link Micro Biotinylation kits are available?

There are four Micro Biotinylation Kits as shown below:

• EZ-Link Micro Sulfo-NHS-Biotinylation Kit, Cat. No. 21925
• EZ-Link Micro Sulfo-NHS-LC-Biotinylation Kit, Cat. No. 21935
• EZ-Link Micro Sulfo-NHS-SS- Biotinylation Kit, Cat. No. 21945
• EZ-Link Micro NHS-PEG4-Biotinylation Kit, Cat. No. 21955

Can I use the HABA assay to verify conjugation after using the EZ-Link Micro Biotinylation Kits?

The HABA assay requires a large volume of biotinylated sample. Using the HABA assay with the Micro Biotinylation kits consumes too much of the sample. An alternative verification method is to perform a dot blot of the biotinylated protein using streptavidin-HRP or other labeled avidin product.

How do the EZ-Link Micro Biotinylation Kits differ from the standard biotinylation kits?

The standard biotinylation kits contain components for labeling 1-10 mg of protein, and the Micro Biotinylation Kits are structured for labeling 50-200 µg of protein in 200-700 mL. The Micro kits contain the biotinylation reagents in small, individual packages and do not include components to perform the HABA assay.

Can you provide the shelf-life for EZ-Link Micro Sulfo-NHS-Biotinylation Kit?

EZ-Link Micro Sulfo-NHS-Biotinylation Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.


For Research Use Only. Not for use in diagnostic procedures.