missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-SS-PEG4-Biotin

Catalog No. PI21442
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21442 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21442 Supplier Thermo Scientific™ Supplier No. 21442

Biotinylate protein amines with this cleavable, long (37.9A) NHS-ester biotinylation reagent containing a reducible disulfide bond in the pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-SS-PEG4-Biotin includes disulfide and polyethylene glycol (PEG) groups in its spacer arm to provide biotinylation that is cleavable and which enhances solubility.

Highlights:

  • Extended spacer arm reduces steric hindrance in binding to avidin, streptavidin and Thermo Scientific NeutrAvidin Protein
  • PEG spacer maintains water solubility of the modified compound
  • Disulfide bond in the spacer arm enables the biotin group to be cleaved from the protein by treatment with reducing agent
  • Useful in affinity purification experiments where it is necessary to recover biotin labeled protein from immobilized avidin, streptavidin or NeutrAvidin™ Supports

Specifications

Product Type NHS-SS-PEG4-Biotin
Quantity 50 mg
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Extra-long, Pegylated, Cleavable
Label Type Biotin & Analogs
Show More Show Less
Can you provide the shelf-life for the EZ-Link NHS-SS-PEG4-Biotin?

The EZ-Link NHS-SS-PEG4-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.