missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-PEG4 Biotinylation Kit

Catalog No. PI21455
Change view
Click to view available options
Quantity:
8 Reaction Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21455 8 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21455 Supplier Thermo Scientific™ Supplier No. 21455

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use

Specifications

Description EZ-Link NHS-PEG4 Biotinylation Kit
Product Type NHS-PEG4 Biotinylation Kit
Quantity 8 Reaction Kit
Content And Storage Sufficient For: 8 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link NHS-PEG4-Biotin, 8 x 2 mg
• PBS Pack (makes 500mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10mg

Store avidin and microtubes of biotin reagent at -20°C. Store PBS at room temperature. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Spacer Pegylated
Labeling Target Antibodies, Proteins
How do the EZ-Link Micro Biotinylation Kits differ from the standard biotinylation kits?

The standard biotinylation kits contain components for labeling 1-10 mg of protein, and the Micro Biotinylation Kits are structured for labeling 50-200 µg of protein in 200-700 mL. The Micro kits contain the biotinylation reagents in small, individual packages and do not include components to perform the HABA assay.

Can you provide the shelf-life for EZ-Link NHS-PEG4 Biotinylation Kit?

EZ-Link NHS-PEG4 Biotinylation Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.