missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-Biotin

Catalog No. PI20217
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20217 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20217 Supplier Thermo Scientific™ Supplier No. 20217
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotinylate protein amines (e.g., lysines) with a simple, short (13.5A), NHS-ester biotinylation reagent containing a membrane-permeable spacer arm for intracellular labeling.

Thermo Scientific™ EZ-Link NHS-Biotin is the simplest NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for general intracellular labeling.

Highlights:

  • Forms stable amide bonds
  • Reacts with primary amines at pH 7-9
  • Must be dissolved in DMSO or DMF and can then be transferred to an aqueous reaction solution
  • Contains a shorter spacer arm than NHS-LC-Biotin
  • Penetrates cell membranes because it is uncharged

Specifications

Product Type NHS-Biotin
Quantity 100 mg
Content And Storage Store desiccated at room temperature.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Short
Label Type Biotin & Analogs
Show More Show Less
Can you provide the shelf-life for EZ-Link NHS-Biotin?

EZ-Link NHS-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (room temperature). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.